Eurotherm AP1080-2000-1 Limit Alarm, DC input, single set point, DPDT; 120/240 VAC, 50-60 Hz
Representative image only.
Eurotherm Mfr # AP1080-2000-1 – Item # EW-68477-98
Field configurable - compact design
PRODUCT OPTIONS
$340.00USD / Each
For better pricing please sign in.
SPECS & DESCRIPTION
Specifications & Description
  • Max Input (VDC)200
  • Min Input (VDC)0.01
  • Input typeDC Current
  • Power Supply (VAC)120/240
  • DescriptionLimit Alarm, DC input, single set point, DPDT
More About this Item
Limit Alarms are setpoint controllers that regulate industrial processes, whether discrete, batch or continuous. Alarms are indicators of problems, be they production line faults, process upsets, equipment failures or control system abnormalities.