Eurotherm AP1290-2000-1 Limit Alarm, thermocouple input, dual set point, SPDT; 120/240 VAC, 50-60 Hz
Representative image only.
Eurotherm Mfr # 1290-2000-1 – Item # EW-68478-00
Field configurable - compact design
PRODUCT OPTIONS
$413.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • Input typeThermocouple
  • Power Supply (VAC)120/240
  • DescriptionLimit Alarm, thermocouple input, dual set point, SPDT
More About this Item
Limit Alarms are setpoint controllers that regulate industrial processes, whether discrete, batch or continuous. Alarms are indicators of problems, be they production line faults, process upsets, equipment failures or control system abnormalities.