Eurotherm Nanodac Recorder/Controller; Loop/LRD/Zirconia/Basic; VAC
Representative image only.
Eurotherm Mfr # Nanodac/VH/C/X/LRD/ZC/TS/XXXX/Basic – Item # EW-70001-43
These units available with optional humidity and sterilizer blocks
  • Option of two independent PID control loops
  • Secure, binary data (UHH) or open (CSV) data files
  • 4 universal input channels with 125 ms parallel sampling
  • 14 additional channels for use as math functions, totalizers or counters
  • 8 Hz sample and recording rate
  • High-density I/O
  • USB and Ethernet ports
  • Simple to configure via push buttons or iTools software
  • Crystal-clear, full-color display
  • Space-saving, 1/4 DIN panel-mount design
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DIN Size0.25
  • Control Input VDCYes
  • Control Input mAYes
  • Control Input RTDYes
  • Control Input ThermocoupleYes
  • Control Output 1Logic
  • Control Output 2Relay
  • Control Output 3DC
  • DescriptionRecorder/Controller; Loop/LRD/Zirconia/Basic; VAC
  • Warranty1 year
1 Year Warranty
Key Features
  • Option of two independent PID control loops
  • Secure, binary data (UHH) or open (CSV) data files
  • 4 universal input channels with 125 ms parallel sampling
  • 14 additional channels for use as math functions, totalizers or counters
  • 8 Hz sample and recording rate
  • High-density I/O
  • USB and Ethernet ports
  • Simple to configure via push buttons or iTools software
  • Crystal-clear, full-color display
  • Space-saving, 1/4 DIN panel-mount design
  • 1 year warranty
More About this Item
The nanodac paperless recorder features secure data recording, graphical display, and optional PID control — all in a compact 1/4 DIN panel-mount design. Full-color 3.5” TFT provides a crystal-clear operator interface for monitoring and process control. Choose to view your process information with horizontal and vertical trends, bar graphs or numeric values, and easily display the alarm status and control loops. Four front-panel push buttons allow simple scrolling between configured views.
Each nanodac contains 50 MB of nonvolatile Flash memory for data storage. All process data is continually logged to this memory and the recorder offers multiple archive strategies to ensure that your data is never lost. You can automatically archive data to a database, to a removable USB memory stick, or over the Ethernet via FTP site.
Recorder functionality is supported by PC-based tools to get data in the way you need it. iTools software creates a secure library for data from which charts can be reviewed, printed and exported to other PC packages as required. All data is encrypted in binary files (UHH format) and can be configured to run over the network which allows for a third data backup route.
Choose models with two independent PIC control loops for reliable control of your process. Functionality includes one of the best autotune facilities available along with overshoot inhibition (cutbacks); compensation for power fluctuations using power feedforward; and linear, fan, oil, and water cooling. Configuration is also done using the downloadable iTools software; any configuration created for a nanodac recorder/controller can be stored, modified offline, and cloned for use in other Eurotherm instruments, saving you time and money.
The optional carbon zirconia block, normally used with control loops, is used for percentage carbon control in furnace atmospheres; the nanodac can also be used as a carbon monitor.
Humidity and Sterilizer Blocks
Sterilizer Application Block
  • Suitible for independent monitoring of sterilizers
  • Up to 4 Input Variables
  • Temperature / Pressure / Conductivity / Flow
  • Indicates Equilibrium time and cycle time
  • Automatic Cycle Number
  • Name files by Cycle No.
  • Sterilization band for each input
  • Cycle Pass / Fail messages
  • F-value (F0 / A0)
    Relative Humidity
  • Relativity Humidity & Dew point
  • Wet & Dry Bulb
  • Altitude compensation
  • Standard I/O for Valve/Solenoid control
  • REVIEWS