Eurotherm T287 Universal Input Transmitter, 4-20 mA Output
Representative image only.
Eurotherm-Action Instruments Mfr # T287-0000 – Item # EW-68478-15
  • PC based user friendly configuration
  • 2000 VDC input to output isolation
  • Single or dual inputs
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Output4 to 20 mA
  • Min Temperature (° C)-40
  • Max Temperature (° C)85
  • DescriptionUniversal Input Transmitter, 4-20 Ma Output
  • Warranty3 Years
3 Year Warranty
Key Features
  • PC based user friendly configuration
  • 2000 VDC input to output isolation
  • Single or dual inputs
  • 3 year warranty
More About this Item
Intelligent, universal process two-wire transmitters feature six types of inputs, DC voltage or current, potentiometer, thermocouple, AC voltage or current, RTD,and
frequency.