Eutech Instruments COND 560 Conductivity Controller with NIST-Traceable Calibration; 110/220 VAC
Representative image only.
Oakton Mfr # 1950540NIST – Item # EW-19505-90
Small size, economical monitor or controller with display
  • Both models measure and display conductivity and temperature
  • Automatic or manual temperature compensation
  • CON 560 has two set points with adjustable hysteresis for on/off control
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Control TypeOn/Off
  • Display TypeLCD
  • Temperature CompensationAutomatic or Manual
  • DescriptionCOND 560 Conductivity Controller, 110/220 VAC, NIST-traceable
  • Warranty1 year
1 Year Warranty
Key Features
  • Both models measure and display conductivity and temperature
  • Automatic or manual temperature compensation
  • CON 560 has two set points with adjustable hysteresis for on/off control
  • 1 year warranty
More About this Item
The CON 550 and 560 monitors are used to measure conductivity and temperature values in applications such as water treatment and monitoring, plating, chemical processing, or food processing. The monitors have user-friendly features like menu-driven programs and calibrations that are easily performed with the push button key pad. All setup and calibration parameters are retained in nonvolatile memory to prevent data loss during power outages.
Measurements can be compensated for temperature variations automatically or manually. Independent calibration and process temperatures can be set when manual temperature compensation is used. All values, operational, and error messages are indicated on the large, dual-line LCD. The units are powered by an universal (110/220 VAC) to 9 VDC power adapter (included). Order 2- or 4-electrode conductivity cells separately.
The CON 560 controller has additional features including two switching contacts as set point relays. The relays have separately adjustable high and low set point hysteresis (dead bands) to prevent relay chattering. The relays feature a 0 to 2000 second time delay to prevent false alarms and unneeded control. The CON 560 also features a backlit and UV light protected display.