Evsco 22-PP-RA4 Two-Way Elliptical Valve, PP, Pneumatic, 1/4"
Representative image only.
Evsco Mfr # 22-PP-RA4 – Item # EW-98154-03
Completely disassemble in-line for easy cleaning
PRODUCT OPTIONS
$296.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.25
  • Outlet Port1/4" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/4" NPT(F)
  • Orifice (in)0.25
  • Orifice (mm)6.35
  • Housing MaterialPP
  • Media CompatibilityUse for liquid, gas, and vacuum applications
  • Actuation MotorPneumatic
  • DescriptionTwo-Way Elliptical Valve, PP, Pneumatic, 1/4"
More About this Item
Use these actuated elliptical valves for liquid, gas, and vacuum applications. Valve systems are available in PP or PVDF bodies; all are FDA compliant and have O-rings made of Viton®. Rated for 150 psi at 73°F (10 bar at 22°C) (liquids).
Pneumatically Actuated Valves are double acting air-to-air and feature 1/4" NPT(F) connection. Control time is 1 second at 40 psi air pressure. Spring return actuators are also available; please call our Application Specialists for details.