This item is now discontinued. Please browse similar items
Evsco 32-PV-RA4 Two-Way Elliptical Valve, PVDF, Pneumatic, 3/8"
Representative image only.
Evsco Mfr # 32-PV-RA4 – Item # EW-98154-22
Completely disassemble in-line for easy cleaning
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.375
  • Outlet Port3/8" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port3/8" NPT(F)
  • Orifice (in)0.375
  • Orifice (mm)9.525
  • Housing MaterialPVDF
  • Media CompatibilityUse for liquid, gas, and vacuumapplications
  • Actuation MotorPneumatic
  • DescriptionTwo-Way Elliptical Valve, PVDF, Pneumatic, 3/8"
More About this Item
Use these actuated elliptical valves for liquid, gas, and vacuum applications. Valve systems are available in PP or PVDF bodies; all are FDA compliant and have O-rings made of Viton®. Rated for 150 psi at 73°F (10 bar at 22°C) (liquids).
Pneumatically Actuated Valves are double acting air-to-air and feature 1/4" NPT(F) connection. Control time is 1 second at 40 psi air pressure. Spring return actuators are also available; please call our Application Specialists for details.