This item is now discontinued. Please browse similar items
Evsco 42-PP-RA4 Two-Way Elliptical Valve, PP, Pneumatic, 1/2"
Representative image only.
Evsco Mfr # 42-PP-RA4 – Item # EW-98154-05
Completely disassemble in-line for easy cleaning
  • FDA-compliant
  • Choose between Polypropylene (PP) or PVDF valve options
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.5
  • Outlet Port1/2" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/2" NPT(F)
  • Orifice (in)0.5
  • Orifice (mm)12.7
  • Housing MaterialPP
  • Media CompatibilityUse for liquid, gas, and vacuumapplications
  • Actuation MotorPneumatic
  • DescriptionTwo-Way Elliptical Valve, PP, Pneumatic, 1/2"
Key Features
  • FDA-compliant
  • Choose between Polypropylene (PP) or PVDF valve options
More About this Item
Use these actuated elliptical valves for liquid, gas, and vacuum applications. Valve systems are available in PP or PVDF bodies; all are FDA compliant and have O-rings made of Viton®. Rated for 150 psi at 73°F (10 bar at 22°C) (liquids).
Pneumatically Actuated Valves are double acting air-to-air and feature 1/4" NPT(F) connection. Control time is 1 second at 40 psi air pressure. Spring return actuators are also available; please call our Application Specialists for details.