Evsco 423-PP-1-RA4 Three-Way Elliptical Valve, PP, Pneumatic, 1/2"
Representative image only.
Evsco Mfr # 423-PP-1-RA4 – Item # EW-98154-15
Completely disassemble in-line for easy cleaning
PRODUCT OPTIONS
$336.10USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.5
  • Outlet Port1/2" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/2" NPT(F)
  • Orifice (in)0.5
  • Orifice (mm)12.7
  • Housing MaterialPP
  • Media CompatibilityUse for liquid, gas, and vacuumapplications
  • Actuation MotorPneumatic
  • DescriptionThree-Way Elliptical Valve, PP, Pneumatic, 1/2"
More About this Item
Use these actuated elliptical valves for liquid, gas, and vacuum applications. Valve systems are available in PP or PVDF bodies; all are FDA compliant and have O-rings made of Viton®. Rated for 150 psi at 73°F (10 bar at 22°C) (liquids).
Pneumatically Actuated Valves are double acting air-to-air and feature 1/4" NPT(F) connection. Control time is 1 second at 40 psi air pressure. Spring return actuators are also available; please call our Application Specialists for details.