Evsco 523-PP-1 PP Three-Way, Two-Position Elliptic Valve, 3/4" NPT(F) connectors
Representative image only.
Evsco Mfr # 523-PP-1 – Item # EW-98150-04
$81.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.75
  • Outlet Port3/4" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port3/4" NPT(F)
  • Orifice (in)0.6875
  • Orifice (mm)17.46
  • Wetted MaterialsPolypropylene, Viton
  • Flow PatternThree-Way, Two-Position
  • DescriptionPP Three-Way, Two-Position Elliptic Valve, 3/4" NPT(F) connectors
More About this Item
Use elliptic valves for liquid, gas, and vacuum applications. Valves are easy to disassemble. Simply pull out the handle and push your stem through the bottom—allowing you to clean and maintain valves in-line. Four-position valves are clearly marked with flow patterns to ensure correct handle position.
Choose from FDA-grade PP or PVDF bodies. All valves have O-rings made of Viton®, 316 SS handles, threaded connections, and nonrestrictive orifices. Rated up to 150 psi at 77°>F and 230°>F (110°>C) at 50 psi (liquids).
ACCESSORIES
Call Us At1-847-549-7600
Live chat