Extech 153117 AC Power Supply Adapter; 115 VAC
Representative image only.
Extech Mfr # 153117 – Item # EW-23402-20
Optimize performance of your Extech meters
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAC Power Supply Adapter; 115 VAC
  • Warranty6 month
6 Month Warranty
Key Features
  • 6 month warranty
More About this Item
Models 20043-01, -02, -03, 23402-20, and -05 are all compatible with the Extech Passive Component LCR meter 20043-00.
Models 20045-59, 64, and -65 are compatible with the Extech MS6000 Series Two-Channel Digital Storage Oscilloscopes 20045-56 and -57.
Meters sold separately.