Extech 365515-BK Stopwatch/Clock with Backlit Display
Representative image only.
Extech Mfr # 365515-BK – Item # EW-35725-09
Won't fail when splashed
  • Water-resistant housing protects watch in harsh environments
  • Stay safe with the 39" (99 cm) L snap-away neck strap
  • Versatile—use as an alarm clock, stopwatch, and calendar
  • Programmable alarm and hourly chime help you stay on task
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Timing Capacity24 hours
  • Timing ModesElapsed timer, split time, two-competitor timer
  • Count DirectionUp
  • ClockYes
  • Resolution0.01 sec up to 30 minutes, 1 sec thereafter
  • Accuracy±3 sec/day
  • AlarmYes
  • DisplayLCD, 1\" H
  • Battery Type1.5 V button cell
  • Battery IncludedYes
  • Height (in)2 3/4
  • Depth (in)1/2
  • Width (in)2 1/4
  • DescriptionStopwatch/Clock with Backlit Display
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Water-resistant housing protects watch in harsh environments
  • Stay safe with the 39" (99 cm) L snap-away neck strap
  • Versatile—use as an alarm clock, stopwatch, and calendar
  • Programmable alarm and hourly chime help you stay on task
  • 1 year warranty
More About this Item
Whether on the plant floor, in a research lab, or outdoors, this combination stopwatch and clock reliably keeps time within easy reach. The water-resistant housing protects the internal components from incidental contact with water. The long neck strap features a snap-away clasp for added safety. Backlight ensures visibility in poor lighting conditions and automatically shuts off after four seconds to conserve battery. Suitable for most timing applications using different modes, including clock, alarm, elapsed timer, split timer, two-competitor timer, and calendar. Programmable alarm with snooze function and hourly chime keeps you from losing track of time.