Extech 801515 SS General Purpose Thermocouple Probe - J Probe
Representative image only.
Extech Mfr # 801515 – Item # EW-95001-82
Choose the type for your unique application
  • Type J/K and RTD available
  • Applications: Alcohols, PCBs, pesticides, pharmaceuticals
  • 1 year warranty
PRODUCT OPTIONS
$45.89USD / Each
$53.99USD / Each
$197.00 - $290.00USD / Each
for an additional
$197.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSs General Purpose Thermocouple Probe - J Probe
  • Warranty1 year
1 Year Warranty
Key Features
  • Type J/K and RTD available
  • Applications: Alcohols, PCBs, pesticides, pharmaceuticals
  • 1 year warranty
More About this Item
Special-application thermocouple probes work with any meter accepting standard thermocouple plug connectors. Choose the Type (J/K) compatible with your meter. Each probe is 39" long.