Extech BR80 Video Borescope Inspection Camera
Representative image only.
Extech Mfr # BR80 – Item # EW-68517-08
17mm camera diameter and 2.4" Color TFT LCD Monitor with lightweight, handheld design to easily find, diagnose, and solve problems.
  • 39" (1 m) flexible gooseneck cable retains configured shape
  • Mini water-proof (IP67) camera head and cable
  • Four bright LED lamps with dimmer to illuminate viewed object
  • Glare-free close-up field of view
  • Vivid 2.4" color LCD monitor for detailed images plus 180° image rotation feature
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionVideo Borescope Inspection Camera
  • Warranty1 year
1 Year Warranty
Key Features
  • 39" (1 m) flexible gooseneck cable retains configured shape
  • Mini water-proof (IP67) camera head and cable
  • Four bright LED lamps with dimmer to illuminate viewed object
  • Glare-free close-up field of view
  • Vivid 2.4" color LCD monitor for detailed images plus 180° image rotation feature