Extech DO700 Multiparameter Meter Kit, includes pH/con/DO electrodes

Representative image only.
Extech Mfr # DO700 – Item # UX-53026-30
Measures DO in mg/L or % saturation, plus 7 other parameters
  • Automatic salinity compensation and manual barometric pressure compensation for increased accuracy
  • Waterproof IP57-rated housing is ideal for field use
  • 1 year warranty

$955.90USD / Each
sign in for your price.

$166.50 - $248.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Include Accessories Today!
Accessories for Extech DO700 Multiparameter Meter
ITEM(S) ON SALE
$87.99 - $110.32USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)19.99
  • pH Resolution0.01/0.1
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 3 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution1 mV
  • Min Conductivity0 to 200 mS
  • Conductivity Resolution0.01 or 0.1 μS; 0.001, 0.01, or 0.1 mS
  • Min TDS0 to 100 ppt
  • TDS Resolution0.01 or 0.1 ppm; 0.001, 0.01, or 0.1 ppt
  • Min Salinity0 to 100 ppt
  • Salinity Resolution0.1 ppt; 0.01%
  • Min Resistivity0 to 100 MΩ
  • Resistivity Resolution1 MΩ
  • Min Dissolved Oxygen0 to 40.00 ppm, 0 to 200%
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature compensationAutomatic
  • WaterproofYes
  • Battery2 AA (included)
  • Width (cm)6.6675
  • Height (cm)12.065
  • Width (in)2 5/8
  • Length (in)1 1/8
  • Height (in)4 3/4
  • DescriptionMultiparameter Meter Kit, includes pH/con/DO electrodes
  • Warranty1 year
1 Year WarrantyConformite EuropeenneISO9001:2000 Certified Supplier

Key Features

  • Automatic salinity compensation and manual barometric pressure compensation for increased accuracy
  • Waterproof IP57-rated housing is ideal for field use
  • 1 year warranty

More About this Item

Versatile meter can read dissolved oxygen concentration/saturation, plus up to 7 other water quality parameters for extreme convenience. Automatic salinity and manual barometric pressure compensation provides accurate DO readings in a variety of conditions. Easy one-button pH calibration can be performed with up to 3 points for better accuracy. One-point conductivity calibration with automatic standard recognition saves time. Additional features include memory for up to 400 readings, large backlit LCD, and auto power off.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat