Extech EA10 Easyview Thermocouple Thermometer - Dual Channel
Representative image only.
Extech Mfr # EA10 – Item # EW-95001-06
Compact and rugged design featuring a large display
  • Accepts any type K thermocouple probe with miniconnector
  • 1 year warranty
PRODUCT OPTIONS
$126.64USD / Each
$148.99USD / Each
$217.50 - $290.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels2
  • Types Of Probes AcceptedType K
  • Min Temperature (° F)-200
  • Max Temperature (° F)1999
  • Min Temperature (° C)-129
  • Max Temperature (° C)1093
  • Accuracy±0.3% of reading +2.0°F (+1.0°C)
  • Resolution0.1°; 1° above 200°
  • Data LoggingNo
  • Display TypeLCD
  • Height (in)6
  • Depth (in)1 1/2
  • No. of Probe(s) Included2
  • Battery Life500 hours
  • Battery6 AAA (included)
  • DescriptionEasyview Thermocouple Thermometer - Dual Channel
  • Warranty1 year
1 Year Warranty
Key Features
  • Accepts any type K thermocouple probe with miniconnector
  • 1 year warranty
More About this Item
These meters offer the functionality to meet any temperature measurement need. Choose the thermometer with the feature that best fits your application.
Model 95001-06 is a dual-input type K thermometer which offers a dual-input type K thermometer with min/max, Hold, °F/°C selection, and auto power off. Plus an offset function for relative measurement, temperature differential between two probes (T1-T2), timer, elapsed time, time of reading, min/max, backlit display, and °F/°C/K selection