Extech EC405 Replacement sensor, conductivity
Representative image only.
Extech Mfr # EC405 – Item # EW-99542-07
Replacement sensor easily snaps into tester
  • Conductivity, TDS, salinity, and temperature measurements
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeAccessory; Probes and Sensors
  • DescriptionReplacement sensor, conductivity
  • Warranty6 month
6 Month Warranty
Key Features
  • Conductivity, TDS, salinity, and temperature measurements
  • 6 month warranty
More About this Item
Sensor is an easy-to-replace module for ExStik II tester. Save by replacing sensor rather than tester.