This item is now discontinued. We recommend the following as suggested replacements: EW-39756-02
Extech MO25 Compact Moisture Pen for Wood and Building Materials
Representative image only.
Extech Mfr # MO25 – Item # EW-39759-04
Ideal for precise, on-the-go measurements
  • Use these pocket-sized moisture meters on wood, drywall, cardboard, plaster, concrete, and mortar
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Moisture Range Wood Scale (Min) (%)5
  • Moisture Range Wood Scale (Max) (%)40
  • Display TypeDigital
  • BatteryOne AAA battery
  • Height (in)1
  • Width (in)51/64
  • Length (in)5 13/32
  • DescriptionCompact Moisture Pen for Wood and Building Materials
  • Warranty1 year
1 Year Warranty
Key Features
  • Use these pocket-sized moisture meters on wood, drywall, cardboard, plaster, concrete, and mortar
  • 1 year warranty
More About this Item
Model MO25 is designed as a quick reference device for determining moisture level in wood and building materials. Five easy-to-read dual color LEDs indicate 10 moisture levels, from 5 to 40% WME. Features include Data Hold, auto off, and low-battery indication.