Extech PQ34-30 Current Clamps, 3000 A, set of 3
Representative image only.
Extech Mfr # PQ34-30 – Item # EW-20041-54
Optimize performance of your Extech meter with these probes
  • Current clamp probes offer different current range options for a wide range of applications
  • Voltage test leads come with alligator clips for accuratley measuring voltage
  • 1 year warranty
PRODUCT OPTIONS
$969.00USD / Each
sign in for your price.
$112.25 - $185.00USD / Each
for an additional
$112.25USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • AC Max (amps)3000
  • DescriptionCurrent Clamps, 3000 A, set of 3
  • Warranty1 Year
1 Year Warranty
Key Features
  • Current clamp probes offer different current range options for a wide range of applications
  • Voltage test leads come with alligator clips for accuratley measuring voltage
  • 1 year warranty
More About this Item
Compatible for use with the Extech three-phase power/harmonics analyzers, models 20046-30 through -33. Meters sold separately.