Eyela NeoDry 7-ES Vacuum Pump; 115 VAC

Representative image only.
Eyela Mfr # 268550 – Item # EW-28625-16
Complete your evaporation system!
  • Vacuum pump
  • 1 year warranty

PRODUCT OPTIONS

$8,593.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Compatible WithEyela Industrial Rotary Evaporators
  • ConnectionsNW25
  • Power (VAC)115
  • Power (Hz)50/60
  • DescriptionVacuum Pump; 115 VAC
  • Warranty1 Year

Key Features

  • Vacuum pump
  • 1 year warranty

More About this Item


Chiller and vacuum pump required for rotary evaporator operation.

Transformer is required for 220 VAC operation. Please contact us for assistance.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat