Filter Cartridge, Pleated Polyester, 5 micron nominal-rated, 9-5/8" L
Representative image only.
Item # EW-29808-21
$116.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyester
  • Filter MediaPolyester
  • Micron Rating5
  • Filter Length (in)9.625
  • Cartridge Length (in)9.625
  • DescriptionFilter Cartridge, Pleated Polyester, 5 micron nominal-rated, 9-5/8" L
More About this Item
Pleated Polyester Cartridges are general purpose cartridges offering long life and high dirt holding capacity. Polyester cartridges are cleanable and reusable in most applications and are NSF 61 listed.