Fischer Technology 602-128 ETA3.3H Eddy Probe, Spring Loaded
Representative image only.
Fischer Technology Mfr # 602-128 – Item # EW-59971-23
Available for sale in U.S., Canada, and Mexico only
  • Easily exchangeable plug-in probes
  • All magnetic induction, eddy current and dual probes can be connected
  • 6 month warranty
$1,003.00USD / Each
$166.50USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Measuring MethodEddy current
  • DisplayLCD
  • ProbeSpring loaded
  • DescriptionETA3.3H Eddy Probe, Spring Loaded
  • Warranty6 month
6 Month Warranty
Key Features
  • Easily exchangeable plug-in probes
  • All magnetic induction, eddy current and dual probes can be connected
  • 6 month warranty
More About this Item
Application-specific probes are the key to successful coating thickness measurement. Probes are the most important component of systems used to measure coating thickness.
Select magnetic induction or eddy current method probes that best meet your application. Fischer probes are adapted and optimized for applications; they are long lasting and reliable.
Probes work with the Fischer Digital Coating Thickness Measurement Dualscope MP20 - which offers an economic solution for the measuring of coatings / platings on ferrous and / or non-ferrous substrate materials.
Probes also work with the Dualscope MP40 - which combines the magnetic induction and eddy current methods in one hand-held instrument, similar to the MP20. The combination enables the user to measure in random order paint, plastic, and organic coatings on non-ferrous metals and on steel, and non-ferrous metal coatings on steel with one and the same instrument.