Fischer Technology F20H Magnetic Induction Probe, Pencil type, 0 to 98.43 mils
Representative image only.
Fischer Technology Mfr # 604-535 – Item # EW-54102-23
Fischer offers a wide range of probes for a variety of measurement tasks
  • Determine heat damage by means of conductivity measurement
  • Determine ferrite content of steel without destruction
  • Available for sale in the United States, Canada, and Mexico only
  • 1 year warranty
PRODUCT OPTIONS
$767.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Range0 to 80 mil (0 to 2000 µm)
  • Measuring MethodMagnetic induction
  • DescriptionMagnetic Induction Probe, Pencil type, 0 to 98.43 mils
  • Warranty1 year
1 Year Warranty
Key Features
  • Determine heat damage by means of conductivity measurement
  • Determine ferrite content of steel without destruction
  • Available for sale in the United States, Canada, and Mexico only
  • 1 year warranty
More About this Item
Choose the appropriate probe for your application.