Fisher BioReagents Amphotericin B Solution, 250μg/mL, 50 mL

Representative image only.
Fisher BioReagents Mfr # BP264550 – Item # EW-88061-43
Reagents and solutions to meet your lab and research needs
  • Amphotericin B is effective against yeast and fungi
  • An antibiotic solution used in cell biology

$150.25USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC47H73NO17
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)50
  • Formula Weight924.1
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number1397-89-3
  • Boiling Point C (° C)100
  • DescriptionAmphotericin B Solution, 250μg/mL, 50 mL
Storage Code Gray

Key Features

  • Amphotericin B is effective against yeast and fungi
  • An antibiotic solution used in cell biology

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat