Fisher BioReagents BP1150500 Isoamyl Alcohol (Molecular Biology) (500mL)

Representative image only.
Fisher BioReagents Mfr # BP1150500 – Item # EW-88056-44

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeMolecular Biology
  • Empirical FormulaC5H12O
  • Linear Formula(CH3)2CHCH2CH2OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • Synonyms3-Methyl-1-butanol; Isopentyl alcohol; Isobutyl carbinol
  • FormLiquid
  • Volume (mL)500
  • Formula Weight88.15
  • PackagingAmber Glass
  • DOT InformationPentanols; Class 3, Packing Group III
  • Storage SymbolRed
  • Hazard SymbolFlammable, Harmful
  • CAS Number123-51-3
  • UN NumberUN1105
  • DescriptionIsoamyl Alcohol (Molecular Biology) (500mL)
FlammableHarmfulStorage Code Red

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat