Fisher BioReagents Tris Base, Molecular Biology Grade, 5 kg

Representative image only.
Fisher BioReagents Mfr # BP1525 – Item # EW-88057-52
Quality and consistency your lab can count on
  • Tris Base is a buffer component in molecular biology, tissue culture, and electrophoresis procedures

$1,353.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeMolecular Biology
  • Empirical FormulaH2NC(CH2OH)3
  • ClassBuffers
  • ChemistryAnalytical Reagents
  • FormSolid
  • SynonymsTrimethylol Aminomethane; Tris Buffer; Tris(hydroxymethyl)aminomethane; Trizma; THAM
  • Weight (g)5000
  • Formula Weight121.14
  • PackagingPoly Pail
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolFlammable, Irritant
  • CAS Number77-86-1
  • Boiling Point C (° C)219
  • Melting Point C (° C)168.5
  • DescriptionTris Base, Molecular Biology Grade, 5 kg
Storage Code Gray

Key Features

  • Tris Base is a buffer component in molecular biology, tissue culture, and electrophoresis procedures

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat