Fisher BioReagents BP1531 Tris Hydrochloride (Small White Flakes/Molecular Biology) (1kg)

Representative image only.
Fisher BioReagents Mfr # BP1531 – Item # EW-88057-53
  • Assay - ≥99%
  • IR - Conforms to standard
  • Melting Point - 152°±2°C
  • Absorbance of a 1M Solution at 280nm - ≤0.05
  • Calcium - ≤10ppm

PRODUCT OPTIONS

$279.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeMolecular Biology
  • Empirical FormulaC4H11NO3•HCl
  • Linear FormulaH2NC(CH2OH)3•HCl
  • ClassBuffers
  • ChemistrySolutions
  • FormFlakes
  • SynonymsTris(hydroxymethyl)aminomethane Hydrochloride; Trizma Hydrochloride
  • Weight (g)1000
  • Formula Weight157.60
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolFlammable, Harmful
  • CAS Number1185-53-1
  • DescriptionTris Hydrochloride (Small White Flakes/Molecular Biology) (1kg)

Key Features

  • Assay - ≥99%
  • IR - Conforms to standard
  • Melting Point - 152°±2°C
  • Absorbance of a 1M Solution at 280nm - ≤0.05
  • Calcium - ≤10ppm
  • Copper - ≤1ppm
  • Iron - ≤2ppm
  • Magnesium - ≤10ppm
  • Lead - ≤5ppm
  • Arsenic - ≤10ppm
  • Barium - ≤1ppm
  • Manganese - ≤1ppm
  • Electrophoresis - To pass test
  • DNase - Not detected
  • RNase - Not detected
  • Protease - Not detected

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat