Fisher BioReagents Bovine Serum Albumin, Heat-Shock Treated, 100 g

Representative image only.
Fisher BioReagents Mfr # BP1600100 – Item # EW-88057-64
Reagents and solutions to meet your lab and research needs
  • Bovine Serum Albumin is used as a protein stabilizer and blocking agent

PRODUCT OPTIONS

$542.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ClassSolids
  • FormSolid
  • SynonymsBSA, Fraction V
  • Weight (g)100
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number9048-46-8
  • DescriptionBovine Serum Albumin, Heat-Shock Treated, 100 g
Storage Code Gray

Key Features

  • Bovine Serum Albumin is used as a protein stabilizer and blocking agent

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat