Fisher BioReagents Sodium Dodecyl Sulfate, Electrophoresis Grade, 500 g

Representative image only.
Fisher BioReagents Mfr # BP166500 – Item # EW-88057-82
Quality and consistency your lab can count on
  • For protein extraction procedures and Sodium Dodecyl Sulfate polyacrylamide gels

$412.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeElectrophoresis
  • Empirical FormulaCH3(CH2)10CH2OSO3Na
  • ChemistryAnalytical Reagents
  • SynonymsSodium Lauryl Sulfate; SDS
  • FormSolid
  • Weight (g)500
  • Formula Weight288.38
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolCorrosive, Flammable, Toxic
  • CAS Number151-21-3
  • Melting Point C (° C)206
  • DescriptionSodium Dodecyl Sulfate, Electrophoresis Grade, 500 g
HarmfulStorage Code Gray

Key Features

  • For protein extraction procedures and Sodium Dodecyl Sulfate polyacrylamide gels

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat