Fisher BioReagents D-Sucrose, Molecular Biology Grade, 10 kg

Representative image only.
Fisher BioReagents Mfr # BP22010 – Item # EW-88058-37
Reagents and solutions to meet your lab and research needs
  • For the preparation of density gradients for DNA ultracentrifugation
  • D-Sucrose is suitable for the preparation of density gradients used in purification of proteins and nucleic acids by ultracentrifugation

$608.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeMolecular Biology
  • Empirical FormulaC10H16N2O3S
  • ClassSolids
  • ChemistryAnalytical Reagents
  • FormSolid
  • Synonymssucrose, saccharose, cane sugar, sugar, table sugar, white sugar
  • Weight (g)10000
  • Formula Weight342.29
  • PackagingPoly Pail
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number57-50-1
  • Melting Point C (° C)190
  • DescriptionD-Sucrose, Molecular Biology Grade, 10 kg
Storage Code Gray

Key Features

  • For the preparation of density gradients for DNA ultracentrifugation
  • D-Sucrose is suitable for the preparation of density gradients used in purification of proteins and nucleic acids by ultracentrifugation

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat