Fisher BioReagents Formamide, Molecular Biology Grade, 500mL

Representative image only.
Fisher BioReagents Mfr # BP227500 – Item # EW-88058-44
Quality and consistency your lab can count on
  • Molecular Biology-grade Formamide is commonly used in nucleic acid hybridization and sequencing

$455.75USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeMolecular Biology
  • Empirical FormulaCH3NO
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Synonymsmethanamide, carbamaldehyde, formimidic acid, formic acid
  • Volume (mL)500
  • Formula Weight45.04
  • PackagingAmber Glass Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolFlammable, Harmful
  • CAS Number75-12-7
  • Boiling Point C (° C)210
  • Melting Point C (° C)2
  • DescriptionFormamide, Molecular Biology Grade, 500mL
HarmfulStorage Code Gray

Key Features

  • Molecular Biology-grade Formamide is commonly used in nucleic acid hybridization and sequencing

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat