Fisher BioReagents BP2291 Glycerol (Molecular Biology) (1 liter) CAS 56-81-5

Representative image only.
Fisher BioReagents Mfr # BP2291 – Item # EW-88058-46

$156.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeMolecular Biology
  • Empirical FormulaC3H8O3
  • Linear FormulaHOCH2CH(OH)CH2OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsGlycerin
  • Volume (mL)1000
  • Formula Weight92.09
  • PackagingGlass bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number56815
  • DescriptionGlycerol (Molecular Biology) (1 liter) CAS 56-81-5

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat