Fisher BioReagents Dimethyl Sulfoxide, 100 mL

Representative image only.
Fisher BioReagents Mfr # BP231100 – Item # EW-88058-50
Quality and consistency your lab can count on
  • DMSO is a cryopreservation agent for cells, and may also be used in bacterial transformations

$107.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical Formula(CH3)2SO
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • SynonymsDMSO
  • Volume (mL)100
  • Formula Weight78.13
  • PackagingGlass Bottle
  • DOT InformationCombustible liquid, n.o.s.; Packing Group III
  • Storage SymbolGray
  • CAS Number67-68-5
  • UN NumberNA1993
  • Boiling Point C (° C)189
  • DescriptionDimethyl Sulfoxide, 100 mL
IrritantStorage Code Gray

Key Features

  • DMSO is a cryopreservation agent for cells, and may also be used in bacterial transformations

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat