Fisher BioReagents D-Biotin, Vitamin B7, 1 g

Representative image only.
Fisher BioReagents Mfr # BP2321 – Item # EW-88058-52
Reagents and solutions to meet your lab and research needs
  • D-Biotin, which forms a stable complex with Avidin, is useful in the study of Avidin-Biotin interactions and in affinity chromatography

$194.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC10H16N2O3S
  • ClassSolids
  • ChemistrySolutions
  • SynonymsVitamin B7
  • FormSolid
  • Weight (g)1
  • Formula Weight244.31
  • PackagingAmber Glass Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number58-85-5
  • Melting Point C (° C)228.9
  • DescriptionD-Biotin, Vitamin B7, 1 g
Storage Code Gray

Key Features

  • D-Biotin, which forms a stable complex with Avidin, is useful in the study of Avidin-Biotin interactions and in affinity chromatography

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat