Fisher BioReagents BP2400500 Acetic Acid, glacial, HPLC grade, 500 mL

Representative image only.
Fisher BioReagents Mfr # BP2400500 – Item # EW-88058-56
Quality and consistency your lab can count on

$38.00USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeHPLC
  • ClassReagent Acids and Bases
  • ChemistryAcids;Inorganics
  • FormLiquid
  • Volume (mL)500
  • DOT InformationAcetic acid, glacial; Class 8, Packing Group III
  • Storage SymbolRed
  • Hazard SymbolCorrosive
  • UN NumberUN2789
  • CAS Number64-19-7
  • DescriptionAcetic Acid, glacial, HPLC grade, (500 mL)

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat