Fisher BioReagents BP2427500 Ammonium Phosphate, Monobasic; 500 g

Representative image only.
Fisher BioReagents Mfr # BP2427500 – Item # EW-88058-86
Reagents and solutions to meet your lab and research needs
  • Assay - ≥98.0%
  • Ammonium Hydroxide Precipitate - ≤0.005%
  • Chloride - ≤5ppm
  • Heavy Metals (as Pb) - ≤5ppm
  • Insoluble Matter - ≤0.005%
  • Iron - ≤0.001%
  • Nitrate - ≤0.001%
  • pH of a 5% Solution at 25°C - 3.8–4.4
  • Potassium - ≤0.005%
  • Sodium - ≤0.005%
  • Sulfur Compounds (as SO4) - ≤0.005%
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaH6NO4P
  • Linear Formula(NH4)H2PO4
  • ClassReagent Salts
  • ChemistrySolutions
  • FormPowder
  • Weight (g)500
  • Formula Weight115.03
  • PackagingGlass bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number7722-76-1
  • DescriptionAmmonium Phosphate (Monobasic) (500g) CAS 7722-76-1
IrritantStorage Code Gray

Key Features

  • Assay - ≥98.0%
  • Ammonium Hydroxide Precipitate - ≤0.005%
  • Chloride - ≤5ppm
  • Heavy Metals (as Pb) - ≤5ppm
  • Insoluble Matter - ≤0.005%
  • Iron - ≤0.001%
  • Nitrate - ≤0.001%
  • pH of a 5% Solution at 25°C - 3.8–4.4
  • Potassium - ≤0.005%
  • Sodium - ≤0.005%
  • Sulfur Compounds (as SO4) - ≤0.005%

More About this Item

Quality and consistency your lab can count on.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat