Fisher BioReagents Phosphate Buffered Saline, 1X Solution, 4 L

Representative image only.
Fisher BioReagents Mfr # BP24384 – Item # EW-88058-95
Reagents and solutions to meet your lab and research needs
  • Filtered and tested for the absence of DNase, RNase, and protease
  • PBS is used in many biochemical applications to maintain pH

PRODUCT OPTIONS

$248.50USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1X
  • ClassBuffers
  • ChemistrySolutions
  • SynonymsPBS
  • FormLiquid
  • Volume (mL)4000
  • PackagingPoly Cube
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number7647-14-5 (Sodium Chloride) 7558-79-4 (Sodium Phosphate Dibasic) 7778-77-0 (Potassium Phosphate Monobasic) 7447-40-7 (Potassium Chloride)
  • DescriptionPhosphate Buffered Saline, 1X Solution, 4 L
Storage Code Gray

Key Features

  • Filtered and tested for the absence of DNase, RNase, and protease
  • PBS is used in many biochemical applications to maintain pH

More About this Item

Choose from powdered or solution forms. Powdered solid form provides a convenient way to make stock PBS solution by just adding water. Solution liquid form simplifies the buffer preparation step by using concentrated solution – simply dilute as needed.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat