Fisher BioReagents BP2466500 2X YT Agar (Powder/Molecular Genetics) (500g)

Representative image only.
Fisher Chemical Mfr # BP2466500 – Item # EW-88059-11
Reagents and solutions to meet your lab and research needs
  • Growth promotion ability:
  • S. aureus - Found
  • E. coli - Found
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeMolecular genetics
  • ClassSolids
  • FormPowder
  • Weight (g)500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Description2X YT Agar (Powder/Molecular Genetics) (500g)
Storage Code Gray

Key Features

  • Growth promotion ability:
  • S. aureus - Found
  • E. coli - Found

More About this Item

Quality and consistency your lab can count on.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat