Fisher BioReagents Water, Sterile, DNA Grade, 1 L

Representative image only.
Fisher BioReagents Mfr # BP24701 – Item # EW-88042-66
Quality and consistency your lab can count on
  • Water of various purity grades suitable for an array of applications
  • DNA-grade Water is DNase- and Protease-free and is suitable for all DNA work

PRODUCT OPTIONS

$125.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeDNA
  • Empirical FormulaH2O
  • FormLiquid
  • Synonymsdihydrogen oxide, dihydrogen monoxide
  • Volume (mL)1000
  • Formula Weight18.02
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7732-18-5
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • DescriptionWater, Sterile, DNA Grade, 1 L
Storage Code Gray

Key Features

  • Water of various purity grades suitable for an array of applications
  • DNA-grade Water is DNase- and Protease-free and is suitable for all DNA work

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat