Fisher BioReagents Tris Buffered Saline Solution, Molecular Biology Grade, 10X, 1 L

Representative image only.
Fisher BioReagents Mfr # BP24711 – Item # EW-88059-24
Reagents and solutions to meet your lab and research needs
  • Save time and simplify your buffer preparation step by using concentrated TBS buffer solution - simply dilute as needed
  • Filtered and autoclaved - ready to use stock TBS buffer
  • Tested for the absence of DNase, RNase, and protease
  • TBS buffer is often used in immunodetection techniques such as Western blotting protocols

PRODUCT OPTIONS

$202.75USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration10X
  • Purity GradeMolecular Biology
  • ClassBuffers
  • ChemistrySolutions
  • FormLiquid
  • SynonymsTBS
  • Volume (mL)1000
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number77-86-1 (Tris) 7447-40-7 (Potassium Chloride) 7647-14-5 (Sodium Chloride) 7647-01-0 (Hydrochloric acid)
  • Boiling Point C (° C)100
  • DescriptionTris Buffered Saline Solution, Molecular Biology Grade, 10X, 1 L
Storage Code Gray

Key Features

  • Save time and simplify your buffer preparation step by using concentrated TBS buffer solution - simply dilute as needed
  • Filtered and autoclaved - ready to use stock TBS buffer
  • Tested for the absence of DNase, RNase, and protease
  • TBS buffer is often used in immunodetection techniques such as Western blotting protocols

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat