Fisher BioReagents Sodium Chloride-Tris-EDTA Buffer, 1X Solution, 1 L

Representative image only.
Fisher Chemical Mfr # BP24781 – Item # EW-88059-45
Reagents and solutions to meet your lab and research needs
  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X STE buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • 1X solution contains 10mM tris, 1mM EDTA, and 100mM NaCl
  • Filtered through a 5 micron filter and autoclaved
  • Tested for the absence of DNase, RNase, and protease
  • STE buffer is used for DNA extraction protocols

SPECS & DESCRIPTION

Specifications & Description

  • ClassBuffers
  • ChemistrySolutions
  • FormLiquid
  • SynonymsSaline Tris–EDTA, TEN
  • Volume (mL)1000
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number60-00-4 (EDTA) 7647-01-0 (HCl) 7647-14-5 (NaCl)
  • Boiling Point C (° C)100
  • DescriptionSodium Chloride-Tris-EDTA Buffer, 1X Solution, 1 L
Storage Code Gray

Key Features

  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X STE buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • 1X solution contains 10mM tris, 1mM EDTA, and 100mM NaCl
  • Filtered through a 5 micron filter and autoclaved
  • Tested for the absence of DNase, RNase, and protease
  • STE buffer is used for DNA extraction protocols

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat