Fisher BioReagents BP250340 Aprotinin (From Bovine Lung) (40 mL) CAS 9087-70-1

Representative image only.
Fisher BioReagents Mfr # BP250340 – Item # EW-88059-71
Reagents and solutions to meet your lab and research needs
  • Aprotinin is competitive serine protease inhibitor that inhibits trypsin, chymotrypsin, kallikrein, plasmin, and other proteolytic enzymes
  • Aprotinin is useful in cell and tissue culture studies to prevent protein degradation
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeAnalytical Reagent
  • ClassSolids
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)40
  • PackagingAmpule
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number9087-70-1
  • DescriptionAprotinin (From Bovine Lung) (40 mL) CAS 9087-70-1
Storage Code Gray

Key Features

  • Aprotinin is competitive serine protease inhibitor that inhibits trypsin, chymotrypsin, kallikrein, plasmin, and other proteolytic enzymes
  • Aprotinin is useful in cell and tissue culture studies to prevent protein degradation

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat