Fisher BioReagents BP2633500 Hydrogen Peroxide (30% in Water) (500g) CAS 7722-84-1

Representative image only.
Fisher BioReagents Mfr # BP2633500 – Item # EW-88061-24
Quality and consistency your lab can count on

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaH2O2
  • Linear FormulaH2O2
  • ClassGeneral Solutions
  • FormLiquid
  • SynonymsHydrogen Dioxide, Hyperoxide
  • Volume (mL)454
  • Weight (g)500
  • Formula Weight34.01
  • PackagingPoly Bottle
  • DOT InformationHYDROGEN PEROXIDE, AQUEOUS SOLUTIONS; Class 5.1 (8), Packing Group II
  • Storage SymbolYellow
  • Hazard SymbolCorrosive, Oxidizing
  • CAS Number7722-84-1 and 7732-18-5
  • UN NumberUN2014
  • DescriptionHydrogen Peroxide (30% in Water) (500 g) CAS 7722-84-1
CorrosiveOxidizing

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat