Fisher BioReagents BP299100 HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology) (100 mL)

Representative image only.
Fisher BioReagents Mfr # BP299100 – Item # EW-88062-82
  • Molarity - 1.0±0.05
  • pH at 37°C - 7.3±0.1
  • Absorbance of a 0.1M Solution at 280nm - ≤0.004
  • &tab; at 260nm - ≤0.008
  • DNase - Not detected

PRODUCT OPTIONS

$104.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeMolecular Biology
  • Empirical FormulaC8H18N2O4S
  • ClassBuffers
  • ChemistrySolutions
  • FormLiquid
  • SynonymsN-(2-Hydroxyethyl)piperazine-N«-2-ethanesulfonic Acid
  • Volume (mL)100
  • Formula Weight238.3
  • PackagingNalgene Poly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number7365-45-9 and 7732-18-5
  • DescriptionHEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology) (100 mL)

Key Features

  • Molarity - 1.0±0.05
  • pH at 37°C - 7.3±0.1
  • Absorbance of a 0.1M Solution at 280nm - ≤0.004
  • &tab; at 260nm - ≤0.008
  • DNase - Not detected
  • RNase - Not detected
  • Protease - Not detected

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat