Fisher BioReagents HEPES, Molecular Biology Grade, 100g

Representative image only.
Fisher BioReagents Mfr # BP310100 – Item # EW-88062-94
Quality and consistency your lab can count on
  • Commonly used buffering agent

$235.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeMolecular Biology
  • Empirical FormulaC8H18N2O4S
  • ClassBuffers
  • ChemistryAnalytical Reagents
  • FormSolid
  • SynonymsN-(2-Hydroxyethyl)piperazine-N«-2-ethanesulfonic Acid
  • Weight (g)100
  • Formula Weight260.28
  • PackagingGlass Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number7365-45-9
  • DescriptionHEPES, Molecular Biology Grade, 100g
Storage Code Gray

Key Features

  • Commonly used buffering agent

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat