Fisher BioReagents BP323100 Sodium Iodide (White Crystals/Molecular Biology) (100g)

Representative image only.
Fisher BioReagents Mfr # BP323100 – Item # EW-88064-02

$138.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeMolecular Biology
  • Empirical FormulaNaI
  • ClassReagent Salts
  • ChemistrySolutions
  • FormCrystalline
  • Weight (g)100
  • Formula Weight149.89
  • PackagingGlass bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number7681-82-5
  • DescriptionSodium Iodide (White Crystals/Molecular Biology) (100g)

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat