Fisher BioReagents Sodium Bicarbonate, 1 kg

Representative image only.
Fisher BioReagents Mfr # BP3281 – Item # EW-88064-09
Quality and consistency your lab can count on
  • This sodium salt is used in buffers for molecular biology and cell culture applications

$124.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaNaHCO3
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • Synonymsbaking soda
  • FormSolid
  • Weight (g)1000
  • Formula Weight84.01
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number144-55-8
  • Melting Point C (° C)270
  • DescriptionSodium Bicarbonate, 1 kg

Key Features

  • This sodium salt is used in buffers for molecular biology and cell culture applications

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat