Fisher BioReagents Sodium Phosphate Dibasic Anhydrous, 500 g

Representative image only.
Fisher BioReagents Mfr # BP332500 – Item # EW-88064-59
Quality and consistency your lab can count on
  • Sodium Phosphate Dibasic Anhydrous (White Granules or Powder)
  • This buffer component is commonly used in tissue culture and molecular biology applications

$156.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaNa2HPO4
  • ClassBuffers
  • ChemistryAnalytical Reagents
  • FormSolid
  • SynonymsDisodium Hydrogen Phosphate
  • Weight (g)500
  • Formula Weight141.96
  • PackagingGlass Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number7558-79-4
  • DescriptionSodium Phosphate Dibasic Anhydrous, 500 g
Storage Code Gray

Key Features

  • Sodium Phosphate Dibasic Anhydrous (White Granules or Powder)
  • This buffer component is commonly used in tissue culture and molecular biology applications

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat