Fisher BioReagents Sodium Tartrate Dihydrate, 500 g

Representative image only.
Fisher BioReagents Mfr # BP352500 – Item # EW-88066-73
Reagents and solutions to meet your lab and research needs
  • This sodium salt is used in buffers for molecular biology and cell culture applications

$364.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaNa2C4H4O6•2H2O
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • SynonymsDisodium Tartrate, Dihydrate
  • FormSolid
  • Weight (g)500
  • Formula Weight230.08
  • PackagingGlass Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number6106-24-7
  • DescriptionSodium Tartrate Dihydrate, 500 g
Storage Code Gray

Key Features

  • This sodium salt is used in buffers for molecular biology and cell culture applications

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat