Fisher BioReagents Sodium Hydroxide, 500 g

Representative image only.
Fisher BioReagents Mfr # BP359500 – Item # EW-88066-83
Quality and consistency your lab can count on
  • Sodium Hydroxide is used to adjust the pH of buffers for molecular biology and cell culture applications

$98.10USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaNaOH
  • ClassReagent Acids and Bases
  • ChemistryAnalytical Reagents
  • FormSolid
  • Synonymscaustic soda
  • Weight (g)500
  • Formula Weight40
  • PackagingPoly Bottle
  • DOT InformationSodium hydroxide, solid; Class 8, Packing Group II
  • Storage SymbolWhite
  • Hazard SymbolCorrosive
  • CAS Number1310-73-2
  • UN NumberUN1823
  • Boiling Point C (° C)1390
  • Melting Point C (° C)318
  • DescriptionSodium Hydroxide, 500 g

Key Features

  • Sodium Hydroxide is used to adjust the pH of buffers for molecular biology and cell culture applications

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat